n-Octadecane 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Octadecane
A986762-1000G
A986762-100G
A986762-25G
A986762-500G
76729-420EA
79.57
USD
76729-420
76729-422
76729-424
76729-426
n-Octadecane 99%
n-Octadecane
Precaución: for Research Use Only
Formula:
H₃C(CH₂)₁₆CH₃ MW: 254.5 g/mol |
MDL Number:
MFCD00009007 CAS Number: 593-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...