N,N-Dimethylacetamide dimethyl acetal 90% stabilised
Supplier: Ambeed
|
Danger
|
Synonyms:
(1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine
A986688-5G
A986688-25G
76729-418EA
8
USD
76729-418
77552-822
N,N-Dimethylacetamide dimethyl acetal 90% stabilised
N,N-Dimethylacetamide dimethyl acetal
Precaución: for Research Use Only
Formula:
C₆H₁₅NO₂ |
MDL Number:
MFCD00008476 CAS Number: 18871-66-4 UN: 1993 ADR: 3,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...