N-(2-Chloro-4-iodophenyl)acetamide 95+%

Supplier: Ambeed
Warning

A985614-1G A985614-25G A985614-5G
76729-412EA 13.8 USD
76729-412 76729-414 76729-416
N-(2-Chloro-4-iodophenyl)acetamide 95+%
N-(2-Chloro-4-iodophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇ClINO
MW: 295.51 g/mol
MDL Number: MFCD02805525
CAS Number: 135050-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR