Moguisteine 99+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(2-((2-methoxyphenoxy)methyl)thiazolidin-3-yl)-3-oxopropanoate, 3-[2-[(2-Methoxyphenoxy)methyl]thiazolidin-3-yl]-3-oxopropanoic acid Ethyl ester, Ethyl 3-[2-[(2-Methoxyphenoxy)methyl]thiazolidin-3-yl]-3-oxopropanoate

A981290-100MG A981290-1G A981290-1MG A981290-250MG
76729-396EA 33.95 USD
76729-396 76729-398 76729-400 76729-402
Moguisteine 99+%
Moguisteine

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₅S
MW: 339.41 g/mol
MDL Number: MFCD00866839
CAS Number: 119637-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR