Palmitic acid N-hydroxysuccinimide ester 98%

Supplier: Ambeed
Warning

A976577-1G A976577-250MG A976577-5G
76729-384EA 31.83 USD
76729-384 76729-386 76729-388
Palmitic acid N-hydroxysuccinimide ester 98%
Palmitic acid N-hydroxysuccinimide ester

Precaución: for Research Use Only

Formula: C₂₀H₃₅NO₄
MW: 353.50 g/mol
MDL Number: MFCD00050402
CAS Number: 14464-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR