p-Tolyl acetate 98%

Supplier: Ambeed
Warning

Synonyms: Acetic acid p-Cresyl ester, Acetic acid p-Tolyl ester, p-Tolyl Acetate, p-Cresyl acetate

A974981-25G A974981-500G A974981-100G
76729-380EA 11.67 USD
76729-380 76729-382 77670-424
p-Tolyl acetate 98%
p-Tolyl acetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.1745 g/mol
MDL Number: MFCD00008703
CAS Number: 140-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR