Potassium-4-(methylthio)phenyltrifluoroborate 95%

Supplier: Ambeed
Warning

A972703-100MG A972703-1G A972703-250MG A972703-5G
76729-372EA 12 USD
76729-372 76729-374 76729-376 76729-378
Potassium-4-(methylthio)phenyltrifluoroborate 95%
Potassium 4-(methylthio)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KS
MDL Number: MFCD07784379
CAS Number: 871231-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR