3-Amino-5-nitrophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A970259-100MG A970259-1G A970259-250MG
76729-354EA 30 USD
76729-354 76729-356 76729-358
3-Amino-5-nitrophenylboronic acid pinacol ester 97%
3-Amino-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BN₂O₄
MDL Number: MFCD07363740
CAS Number: 871329-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR