Octylphosphonic acid 98%

Supplier: Ambeed
Danger

A969311-25G A969311-1G A969311-5G
76729-352EA 40 USD
76729-352 77670-408 77880-788
Octylphosphonic acid 98%
Octylphosphonic acid

Precaución: for Research Use Only

Formula: C₈H₁₉O₃P
MDL Number: MFCD00015841
CAS Number: 4724-48-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR