Methyl 4-(1-hydroxyethyl)benzoate 95%

Supplier: Ambeed
Warning

Synonyms: 1-[4-(Methoxycarbonyl)phenyl]ethan-1-ol, 4-(Methoxycarbonyl)-α-methylbenzyl alcohol

A968955-10G A968955-250MG
76729-348EA 658.82 USD
76729-348 76729-350
Methyl 4-(1-hydroxyethyl)benzoate 95%
Methyl 4-(1-hydroxyethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.203 g/mol
MDL Number: MFCD00216476
CAS Number: 79322-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR