Nickel(II) hydroxide 99%

Supplier: Ambeed
Danger

A962316-25G A962316-100G
76729-340EA 10 USD
76729-340 77603-364
Nickel(II) hydroxide 99%
Nickel(II) hydroxide

Precaución: for Research Use Only

Formula: H₂NiO₂
MDL Number: MFCD00011140
CAS Number: 12054-48-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR