N,N-Dimethyl-N-dodecylbetaine 30% in water

Supplier: Ambeed
Danger

Synonyms: 2-(Dodecyldimethylammonio)acetate

A952537-100G A952537-500G
76729-318EA 10 USD
76729-318 76729-320 76729-322
N,N-Dimethyl-N-dodecylbetaine 30% in water
N,N-Dimethyl-N-dodecylbetaine

Precaución: for Research Use Only

Formula: C₁₆H₃₃NO₂
MDL Number: MFCD00084742
CAS Number: 683-10-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR