Pyrrolidine-1-carbonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 1-Cyanopyrrolidine

A951948-1G A951948-25G A951948-5G
76729-312EA 15.92 USD
76729-312 76729-314 76729-316
Pyrrolidine-1-carbonitrile 98%
Pyrrolidine-1-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₈N₂
MW: 96.13 g/mol
MDL Number: MFCD00003165
CAS Number: 1530-88-7
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR