2-Benzyloxy-5-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Benzyloxy)-5-fluorophenyl)boronic acid, 2-Benzyloxy-5-fluorobenzeneboronic acid

A949987-10G A949987-1G A949987-5G
76729-300EA 109.28 USD
76729-300 76729-302 76729-304
2-Benzyloxy-5-fluorophenylboronic acid 98%
2-Benzyloxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MW: 246.0420 g/mol
MDL Number: MFCD03788403
CAS Number: 779331-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR