Scandium(III) acetate hydrate 98+%

Supplier: Ambeed
Warning

Synonyms: Acetic acid scandium(III) salt hydrate

A947466-1G A947466-5G
76729-294EA 24.41 USD
76729-294 76729-296
Scandium(III) acetate hydrate 98+%
Scandium(III) acetate hydrate

Precaución: for Research Use Only

Formula: C₆H₉O₆Sc·H₂O
MW: 240.10 g/mol
MDL Number: MFCD00150122
CAS Number: 304675-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR