m-Toluidine hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: M-toluidine hydrochloride

A945471-25G A945471-5G
76729-290EA 38 USD
76729-290 76729-292
m-Toluidine hydrochloride 95%
m-Toluidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉N
MDL Number: MFCD00070607
CAS Number: 638-03-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR