tert-Butyl 2-hydroxypiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A945082-100MG A945082-250MG
76729-286EA 87 USD
76729-286 76729-288
tert-Butyl 2-hydroxypiperidine-1-carboxylate 98%
tert-Butyl 2-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD09031186
CAS Number: 86953-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR