Acetyl-d-alloisoleucine 95%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-D-allo-isoleucine

A943077-100MG A943077-250MG
76729-276EA 21 USD
76729-276 76729-278
Acetyl-d-alloisoleucine 95%
Acetyl-d-alloisoleucine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
MDL Number: MFCD00136538
CAS Number: 54831-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR