1,3-Dibutyl-2-thiourea 97%

Supplier: Ambeed
Warning

Synonyms: N,N'-Dibutylthiourea, 1,3-Dibutylthiourea, N,N'-Di-n-butylthiourea

A940299-1000G A940299-100G A940299-500G A940299-25G
76729-270EA 162 USD
76729-270 76729-272 76729-274 77880-761
1,3-Dibutyl-2-thiourea 97%
1,3-Dibutyl-2-thiourea

Precaución: for Research Use Only

Formula: H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃
MW: 188.34 g/mol
Boiling Pt: 252
MDL Number: MFCD00004926
CAS Number: 109-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR