Boc-Phe-OBzl 97%

Supplier: Ambeed
Warning

A931260-1G A931260-25G A931260-5G
76729-224EA 9 USD
76729-224 76729-226 76729-228
Boc-Phe-OBzl 97%

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO₄
MDL Number: MFCD00237576
CAS Number: 66617-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR