1,3-Bis(2-chloroethyl)urea 95%

Supplier: Ambeed
Warning

A922285-1G A922285-5G
76729-196EA 44.56 USD
76729-196 76729-198
1,3-Bis(2-chloroethyl)urea 95%
1,3-Bis(2-chloroethyl)urea

Precaución: for Research Use Only

Formula: C₅H₁₀Cl₂N₂O
MW: 185.052 g/mol
MDL Number: MFCD00068573
CAS Number: 2214-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR