dibenzo[b,d]furan-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: dibenzo[b,d]furan-2-carbaldehyde, Dibenzofuran-2-carboxaldehyde

A917690-1G A917690-250MG A917690-25G A917690-5G
76729-188EA 53.05 USD
76729-188 76729-190 76729-192 76729-194
dibenzo[b,d]furan-2-carbaldehyde 98%
dibenzo[b,d]furan-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈O₂
MW: 196.205 g/mol
MDL Number: MFCD00136272
CAS Number: 5397-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR