Benzyl 4-(3-methoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A913395-1G
A913395-5G
76729-168EA
87
USD
76729-168
77573-832
Benzyl 4-(3-methoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 95%
Benzyl 4-(3-methoxy-3-oxopropyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₈H₂₁NO₄ |
MDL Number:
MFCD00010681 CAS Number: 20303-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...