Pentaerythritol tetrabenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 2,2-Bis((benzoyloxy)methyl)propane-1,3-diyl dibenzoate

A911793-100G A911793-25G A911793-500G A911793-5G
76729-160EA 63.66 USD
76729-160 76729-162 76729-164 76729-166
Pentaerythritol tetrabenzoate 97%
Pentaerythritol tetrabenzoate

Precaución: for Research Use Only

Formula: C₃₃H₂₈O₈
MW: 552.58 g/mol
MDL Number: MFCD00020676
CAS Number: 4196-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR