1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-Cbz-piperidine-3-carboxylate

A909188-10G A909188-5G A909188-100G
76729-156EA 14.86 USD
76729-156 76729-158 77603-272
1-Benzyl 3-methyl piperidine-1,3-dicarboxylate 98%
1-Benzyl 3-methyl piperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₄
MW: 277.316 g/mol
MDL Number: MFCD06795953
CAS Number: 174543-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR