Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I) 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Palladium(I) tri-tert-butylphosphine bromide dimer, Di-mu-bromobis(tri-t-butylphosphino)dipalladium(I)
A908546-100MG
A908546-1G
A908546-250MG
76729-150EA
21
USD
76729-150
76729-152
76729-154
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I) 98%
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Precaución: for Research Use Only
Formula:
C₂₄H₅₆Br₂P₂Pd₂ MW: 779.3 g/mol |
MDL Number:
MFCD04114019 CAS Number: 185812-86-6 UN: 2921 ADR: 8,4.1 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...