Rhodium(II) octanoate dimer 98%

Supplier: Ambeed
Warning

A907319-250MG A907319-100MG
76729-148EA 88 USD
76729-148 77603-268
Rhodium(II) octanoate dimer 98%
Rhodium(II) octanoate dimer

Precaución: for Research Use Only

Formula: C₃₂H₆₀O₈Rh₂
MW: 778.63 g/mol
MDL Number: MFCD00064724
CAS Number: 73482-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR