2-(Phenylthio)nicotinic acid 97%

Supplier: Ambeed
Warning

A904354-1G A904354-250MG A904354-5G
76729-124EA 24.41 USD
76729-124 76729-126 76729-128 76729-130
2-(Phenylthio)nicotinic acid 97%
2-(Phenylthio)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂S
MDL Number: MFCD00052110
CAS Number: 35620-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR