Piroctone olamine 97%

Supplier: Ambeed
Danger

Synonyms: 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine, 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2(1H)-one 2-aminoethanol salt

A903889-100MG A903889-50MG A903889-1MG
76729-120EA 9 USD
76729-120 76729-122 77670-330
Piroctone olamine 97%
Piroctone olamine

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₂
MW: 298.43 g/mol
MDL Number: MFCD01690792
CAS Number: 68890-66-4
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR