Palmitic acid hydrazide 98% (by ELSD)
Supplier: Ambeed
|
Danger
|
Synonyms:
Palmitohydrazide
A901002-1G
A901002-25G
A901002-5G
76729-116EA
11
USD
76729-116
76729-118
77670-322
Palmitic acid hydrazide 98% (by ELSD)
Palmitic acid hydrazide
Precaución: for Research Use Only
Formula:
C₁₆H₃₄N₂O |
MDL Number:
MFCD00066357 CAS Number: 2619-88-7 UN: 1325 ADR: 4.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...