Diethyl (4-methylbenzyl)phosphonate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 4-methylbenzylphosphonate

A900687-100G A900687-1G A900687-25G A900687-5G
76729-102EA 347.98 USD
76729-102 76729-104 76729-106 76729-108
Diethyl (4-methylbenzyl)phosphonate 97%
Diethyl (4-methylbenzyl)phosphonate

Precaución: for Research Use Only

Formula: C₁₂H₁₉O₃P
MW: 242.26 g/mol
MDL Number: MFCD00041385
CAS Number: 3762-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR