2'-Amino-4'-methoxyacetanilide 95%

Supplier: Ambeed
Warning

A900434-100MG A900434-1G A900434-250MG
76729-096EA 58.35 USD
76729-096 76729-098 76729-100
2'-Amino-4'-methoxyacetanilide 95%
2'-Amino-4'-methoxyacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
MDL Number: MFCD00026124
CAS Number: 5472-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR