3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, 3-aminomethylphenylboronic acid, pinacol ester hydrochloride

A898168-1G A898168-250MG A898168-5G
76729-090EA 59 USD
76729-090 76729-092 76729-094
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride 98%
3-(Aminomethyl)phenylboronic acid pinacol ester, hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₁BClNO₂
MW: 269.58 g/mol
MDL Number: MFCD03410294
CAS Number: 380430-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR