Methyl 4-(piperazin-1-yl)benzoate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-piperazin-1-ylbenzoate

A897458-250MG A897458-1G A897458-5G
76729-088EA 15.92 USD
76729-088 77552-686 77603-242
Methyl 4-(piperazin-1-yl)benzoate 96%
Methyl 4-(piperazin-1-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MW: 220.26 g/mol
MDL Number: MFCD08234800
CAS Number: 163210-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR