Ethyl-4-(butylamino)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Butylamino)benzoic Acid Ethyl Ester, Ethyl 4-(butylamino)benzoate

A896982-1000G A896982-100G A896982-500G A896982-25G
76729-070EA 259.93 USD
76729-070 76729-072 76729-074 77603-240
Ethyl-4-(butylamino)benzoate 98%
Ethyl-4-(butylamino)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 221.30 g/mol
MDL Number: MFCD00017283
CAS Number: 94-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR