N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide 95%

Supplier: Ambeed
Warning

A894958-100MG A894958-1G A894958-250MG
76729-064EA 46 USD
76729-064 76729-066 76729-068
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide 95%
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O
MDL Number: MFCD08529471
CAS Number: 36321-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR