Nifuroxazide 98%

Supplier: Ambeed
Warning

A887401-100G A887401-1G A887401-25G A887401-50MG A887401-1000G A887401-1MG A887401-500G A887401-5G A887401-250MG
76729-044EA 165 USD
76729-044 76729-046 76729-048 76729-050 77670-292 77670-294 77670-296 77670-298 77880-709
Nifuroxazide 98%
Nifuroxazide

Precaución: for Research Use Only

Formula: C₁₂H₉N₃O₅
MDL Number: MFCD00079482
CAS Number: 965-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR