1-(Toluene-4-sulfonyl)indole-3-boronic acid 97%

Supplier: Ambeed
Warning

A887299-100MG A887299-1G A887299-250MG
76729-038EA 40 USD
76729-038 76729-040 76729-042
1-(Toluene-4-sulfonyl)indole-3-boronic acid 97%
1-(Toluene-4-sulphonyl)indole-3-boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄BNO₄S
MW: 315.15 g/mol
MDL Number: MFCD03093101
CAS Number: 149108-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR