Phenyl carbamate 95%

Supplier: Ambeed
Warning

A886104-1G A886104-25G A886104-5G A886104-100G A886104-10G
76729-032EA 9 USD
76729-032 76729-034 76729-036 77880-706 77880-707
Phenyl carbamate 95%
Phenyl carbamate

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MDL Number: MFCD00007961
CAS Number: 622-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR