Methyl-3,5-dibromo-4-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,5-dibromo-4-methylbenzoate

A882597-1G A882597-25G A882597-5G
76729-018EA 16.98 USD
76729-018 76729-020 76729-022
Methyl-3,5-dibromo-4-methylbenzoate 98%
Methyl 3,5-dibromo-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₂
MW: 307.98 g/mol
MDL Number: MFCD00010729
CAS Number: 74896-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR