3-(1-(tert-Butoxycarbonyl)piperidin-3-yl)propanoic acid 95%
Supplier: Ambeed
|
Warning
|
A881900-100MG
A881900-1G
A881900-250MG
A881900-5G
76729-010EA
37.14
USD
76729-010
76729-012
76729-014
76729-016
3-(1-(tert-Butoxycarbonyl)piperidin-3-yl)propanoic acid 95%
3-(1-(tert-Butoxycarbonyl)piperidin-3-yl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₃H₂₃NO₄ MW: 257.32 g/mol |
MDL Number:
MFCD02179008 CAS Number: 352004-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...