3-(4-Morpholinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A879012-100MG A879012-1G A879012-250MG A879012-5G
76728-992EA 20 USD
76728-992 76728-994 76728-996 76728-998
3-(4-Morpholinylcarbonyl)phenylboronic acid 98%
3-(4-Morpholinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BNO₄
MDL Number: MFCD04115687
CAS Number: 723281-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR