Potassium osmate(VI) dihydrate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Potassium osmium(VI) oxide dihydrate, Potassium dioxidodioxoosmium dihydrate, Dipotassium osmate dihydrate
A877216-10G
A877216-1G
A877216-5G
76728-986EA
438.16
USD
76728-986
76728-988
76728-990
Potassium osmate(VI) dihydrate 97%
Potassium osmate(VI) dihydrate
Precaución: for Research Use Only
Formula:
K₂O₄Os·2H₂O MW: 368.45 g/mol |
MDL Number:
MFCD00149919 CAS Number: 10022-66-9 UN: 3085 ADR: 5.1,8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...