Ethyl-4-hydroxy-7-methoxyquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxy-7-methoxyquinoline-3-carboxylate

A875527-1G A875527-25G A875527-5G
76728-980EA 17 USD
76728-980 76728-982 76728-984
Ethyl-4-hydroxy-7-methoxyquinoline-3-carboxylate 98%
Ethyl 4-hydroxy-7-methoxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₄
MDL Number: MFCD00085986
CAS Number: 63463-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR