Pyrido[2,3-d]pyrimidine-2,4(1h,3h)-dione 95%

Supplier: Ambeed
Warning

Synonyms: Pyrido[2,3-d]pyrimidine-2,4(1H,3H)-dione

A874599-1G A874599-5G
76728-962EA 33.95 USD
76728-962 76728-964
Pyrido[2,3-d]pyrimidine-2,4(1h,3h)-dione 95%
Pyrido[2,3-d]pyrimidine-2,4(1h,3h)-dione

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.134 g/mol
MDL Number: MFCD08234704
CAS Number: 21038-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR