4-(4-Formyl-3-methoxyphenoxy)butanoic acid 98%

Supplier: Ambeed
Warning

A872054-25G A872054-5G
76728-958EA 411.63 USD
76728-958 76728-960
4-(4-Formyl-3-methoxyphenoxy)butanoic acid 98%
4-(4-Formyl-3-methoxyphenoxy)butanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₅
MW: 238.23 g/mol
MDL Number: MFCD01317807
CAS Number: 309964-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR