1,3,3-Trimethyl-2-(formylmethylene)indoline 95%

Supplier: Ambeed
Warning

A871268-100G A871268-10G A871268-25G A871268-500G A871268-5G
76728-948EA 137.92 USD
76728-948 76728-950 76728-952 76728-954 76728-956
1,3,3-Trimethyl-2-(formylmethylene)indoline 95%
1,3,3-Trimethyl-2-(formylmethylene)indoline

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO
MW: 201.26 g/mol
MDL Number: MFCD00071813
CAS Number: 84-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR