2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)benzoic acid

A869899-1G A869899-25G A869899-5G
76728-942EA 36 USD
76728-942 76728-944 76728-946
2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid 95+%
2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₉NO₄
MW: 267.24 g/mol
MDL Number: MFCD00041218
CAS Number: 41513-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR