Methyldiphenylphosphine 98%

Supplier: Ambeed
Warning

A868180-100G A868180-10G A868180-1G A868180-25G A868180-5G A868180-500G
76728-930EA 392.54 USD
76728-930 76728-932 76728-934 76728-936 76728-938 77670-260
Methyldiphenylphosphine 98%
Methyldiphenylphosphine

Precaución: for Research Use Only

Formula: C₁₃H₁₃P
MDL Number: MFCD00008508
CAS Number: 1486-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR