Nordihydroguaiaretic acid 98%

Supplier: Ambeed
Warning

A867405-100MG A867405-1MG A867405-25MG
76728-924EA 37.14 USD
76728-924 76728-926 76728-928
Nordihydroguaiaretic acid 98%
Nordihydroguaiaretic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₂O₄
MW: 302.37 g/mol
MDL Number: MFCD00002206
CAS Number: 500-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR